Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, ACS grade, 99.0+%, Thermo Scientific Chemicals

Catalog number: 043894.K7
4 L, Each
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, ACS grade, 99.0+%, Thermo Scientific Chemicals

Catalog number: 043894.K7
4 L, Each
Quantity
Catalog number: 043894.K7
also known as 043894-K7
Price (USD)
Have Questions?

Chemical Identifiers

CAS
872-50-4
IUPAC Name
1-methylpyrrolidin-2-one
Molecular Formula
C5H9NO
InChI Key
SECXISVLQFMRJM-UHFFFAOYSA-N
SMILES
CN1CCCC1=O
Form
Clear Liquid
Assay (GC)
>/= 99.0%
Impurity content
≤ 0.01% Free amines (as CH₃NH₂)
Color (APHA)
≤50
Chloride content
≤ 1ppm

Description

1-methyl-2-pyrrolidinone, is useful for spectrophotometry, chromatography and ICP-MS detection. It is useful for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. It is also useful for pharmaceutical and different industrial applications due to its broad solvency and low volatility.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text