Thermo Scientific Chemicals

Polyethyleneimine, linear, M.W. 25,000, Thermo Scientific Chemicals

Catalog number: 043896.03
1 g, Each
Thermo Scientific Chemicals

Polyethyleneimine, linear, M.W. 25,000, Thermo Scientific Chemicals

Catalog number: 043896.03
1 g, Each
Quantity
Catalog number: 043896.03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9002-98-6
Molecular Formula
((C2H5N)zC2H4N)y(C2H5N)x
SMILES
*-CCNCCN(-*)CCN-*
MDL Number
MFCD00084427
Appearance (Color)
White to yellow
Form
Amorphous to crystalline solid
Comment
Transfection Performance Testing (IgG Expression in HEK293): ≥ 6.5 mg/L
Solution Test
Clear solution- Passes test
Viscosity
Inherent viscosity: 0.22 - 0.40 dL/g

Description

Polyethyleneimine, linear, M.W. 25,000 is used to improve the appearance of negatively charged dyes by modulating their properties and also improving their adherence to surfaces. It binds to DNA and other negatively charges biological molecules, thereby making it for efficient vector carriers in research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text