Ammonium cobalt(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals
Ammonium cobalt(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium cobalt(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals

As spectroscopic standard, cobalt plating, catalysis, ceramics
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 043902.36
also known as 043902-36
Price (USD)/ Each
522.65
Online exclusive
581.00 
Save 58.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
522.65
Online exclusive
581.00 
Save 58.35 (10%)
Add to cart
Ammonium cobalt(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals
Catalog number043902.36
Price (USD)/ Each
522.65
Online exclusive
581.00 
Save 58.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13586-38-4
SpecificationsSpecification SheetSpecification Sheet
Melting Point120-124
Appearance (Color)Pink to Red
Assay (unspecified)99% min
FormPowder or Crystals
Ammonium cobalt(II) sulfate hexahydrate is uses as spectroscopic standard, cobalt plating, catalysis and ceramics. It is also used in pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only