Thermo Scientific Chemicals

(Aminoethylaminomethyl)phenethyltrimethoxysilane, mixture of m and p isomers, Thermo Scientific Chemicals

Catalog number: 043904.04
2 g, Each
Thermo Scientific Chemicals

(Aminoethylaminomethyl)phenethyltrimethoxysilane, mixture of m and p isomers, Thermo Scientific Chemicals

Catalog number: 043904.04
2 g, Each
Quantity
Catalog number: 043904.04
also known as 043904-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
74113-77-2
IUPAC Name
3,3-bis(2,2,6,6-tetramethylpiperidin-4-yl) 1,5-dioxospiro[5.5]undeca-7,9-diene-3,3-dicarboxylate
Molecular Formula
C31H46N2O6
InChI Key
IIZYNJMWYJKDJN-UHFFFAOYSA-N
SMILES
CC1(C)CC(CC(C)(C)N1)OC(=O)C1(CC(=O)C2(CC=CC=C2)C(=O)C1)C(=O)OC1CC(C)(C)NC(C)(C)C1
Assay from Suppliers CofA
≥88% (all isomers)
Appearance (Color)
Pale Yellow
Form
Liquid

Description

As a crosslinking agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text