Chemical Name or MaterialPlatinum
Name Note5% on activated carbon powder, Sulfided, 0.5% S (as sulfide)
CAS7440-6-6
Health Hazard 1H228
Health Hazard 2GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more