Thermo Scientific Chemicals

Cobalt(II) sulfamate hydrate, Thermo Scientific Chemicals

Catalog number: 043906.22
100 g, Each
Thermo Scientific Chemicals

Cobalt(II) sulfamate hydrate, Thermo Scientific Chemicals

Catalog number: 043906.22
100 g, Each
Quantity
Catalog number: 043906.22
also known as 043906-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14017-41-5
IUPAC Name
λ²-cobalt(2+) disulfamate
Molecular Formula
CoH4N2O6S2
InChI Key
WLQXLCXXAPYDIU-UHFFFAOYSA-L
SMILES
[Co++].NS([O-])(=O)=O.NS([O-])(=O)=O
Appearance (Color)
Red
Form
Powder
Assay (special)
Cobalt %: Report Value
Water Content
Report Value

Description

Cobalt(II) sulfamate hydrate is used in photonic device fabrication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text