Thermo Scientific Chemicals

Potassium metabisulfite, 95+%, Thermo Scientific Chemicals

Catalog number: 043908.36
500 g, Each
Thermo Scientific Chemicals

Potassium metabisulfite, 95+%, Thermo Scientific Chemicals

Catalog number: 043908.36
500 g, Each
Quantity
Catalog number: 043908.36
also known as 043908-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16731-55-8
IUPAC Name
dipotassium sulfinatosulfonate
Molecular Formula
K2O5S2
InChI Key
RWPGFSMJFRPDDP-UHFFFAOYSA-L
SMILES
[K+].[K+].[O-]S(=O)S([O-])(=O)=O
Assay (unspecified)
95% min

Description

It serves as a component of certain photographic developers and its solution is used in photographic fixing, in the textile industry for dyeing and cotton printing. It is utilized to precipitate gold from solution in aqua regia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text