4-tert-Butylphenylacetylene, 90+%, Thermo Scientific Chemicals
4-tert-Butylphenylacetylene, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-tert-Butylphenylacetylene, 90+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 043920.04
also known as 043920-04
Price (USD)/ Each
36.40
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
36.40
Add to cart
4-tert-Butylphenylacetylene, 90+%, Thermo Scientific Chemicals
Catalog number043920.04
Price (USD)/ Each
36.40
-
Add to cart
Specifications
Chemical Name or Material4-tert-Butylphenylacetylene
CAS772-38-3
Health Hazard 1H227-H315-H319-H335
Health Hazard 2GHS H Statement
H227
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
4-tert-Butylphenylacetylene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only