Phenyl propargyl ether, 97%, Thermo Scientific Chemicals
Phenyl propargyl ether, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenyl propargyl ether, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 043923.03
also known as 043923-03
Price (USD)/ Each
23.10
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
23.10
Add to cart
Phenyl propargyl ether, 97%, Thermo Scientific Chemicals
Catalog number043923.03
Price (USD)/ Each
23.10
-
Add to cart
Chemical Identifiers
CAS13610-02-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
FormLiquid
Assay from Suppliers CofA≥96.0%
Phenyl propargyl ether is used as building block for synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only