N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 043928.06
also known as 043928-06
Price (USD)/ Each
44.65
Online exclusive
49.20 
Save 4.55 (9%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
44.65
Online exclusive
49.20 
Save 4.55 (9%)
Add to cart
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Catalog number043928.06
Price (USD)/ Each
44.65
Online exclusive
49.20 
Save 4.55 (9%)
-
Add to cart
Chemical Identifiers
CAS25561-30-2
IUPAC Nametrimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)ethanimidate
Molecular FormulaC8H18F3NOSi2
InChI KeyXCOBLONWWXQEBS-UHFFFAOYSA-N
SMILESC[Si](C)(C)OC(=N[Si](C)(C)C)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)10, max
Assay (unspecified)0.5% min. TMCS
Assay (unspecified)93.0% min.
FormClear liquid
N,O-Bis(trimethylsilyl)trifluoroacetamide is used for analytical purposes or as a chemical reagent for synthesis of more complex molecules. It is silylation reagent useful in GC and GC-MS for the derivatization of a wide range of functional groups under mild conditions. It is also used as DAT inhibitor and a SLC6A2 inhibitor. It is routinely used to derivatize carboxylic acids, phenols, steroids, amines, alcohols, and alkaloids and amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only