Thermo Scientific Chemicals

N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS, Thermo Scientific Chemicals

Catalog number: 043939.22
100 g, Each
Thermo Scientific Chemicals

N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS, Thermo Scientific Chemicals

Catalog number: 043939.22
100 g, Each
Quantity
Catalog number: 043939.22
also known as 043939-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25561-30-2
IUPAC Name
trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)ethanimidate
Molecular Formula
C8H18F3NOSi2
InChI Key
XCOBLONWWXQEBS-UHFFFAOYSA-N
SMILES
C[Si](C)(C)OC(=N[Si](C)(C)C)C(F)(F)F
Form
Liquid
Color (APHA)
≤10
Assay (GC)
0.5-4.0% TMCS
Assay (GC)
≥93.0%
Appearance (Color)
Clear

Description

N,O-Bis(trimethylsilyl)trifluoroacetamide is used as a derivatization reagent for gas chromatography and mass-spectrometry analysis of phenolic acids in fruits. It is also used as a reactant in the preparation of pyrimidinone ribosides and analogs which shows anti-tumor properties. It plays as an important role as a reagent for the regioselective desulfation of polysaccharide sulfates. Furthermore, it is used to prepare trimethylsilyl ethers of carbohydrates and alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text