Polyethylene, UHMW, Thermo Scientific Chemicals
Polyethylene, UHMW, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyethylene, UHMW, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Catalog number 043951.30
also known as 043951-30
Price (USD)/ Each
139.65
Online exclusive
155.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
139.65
Online exclusive
155.00 
Save 15.35 (10%)
Add to cart
Polyethylene, UHMW, Thermo Scientific Chemicals
Catalog number043951.30
Price (USD)/ Each
139.65
Online exclusive
155.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9002-88-4
Molecular Formula(C2H4)n
SMILES*-CC-*
MDL NumberMFCD00084423
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Particle Sizemean particle size 150 micron
FormPowder
Polyethylene, UHMW fibers is used in armor, especially personal armor, vehicle armor, climbing equipments, fishing line, bow strings and cut-resistant gloves. Its sheet is used as synthetic ice in ice rinks . In medical field, it is considered as a vital biomaterial used in hip, knee, for spine implants. It is also used in the production of polyvinyl chloride windows and doors, hydraulic seals and bearings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only