1-Aminopyrene, 97%, Thermo Scientific Chemicals
1-Aminopyrene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Aminopyrene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 043968.02
also known as 043968-02
Price (USD)/ Each
49.10
-
Add to cart
Quantity:
0.5 g
Request bulk or custom format
Price (USD)/ Each
49.10
Add to cart
1-Aminopyrene, 97%, Thermo Scientific Chemicals
Catalog number043968.02
Price (USD)/ Each
49.10
-
Add to cart
Chemical Identifiers
CAS1606-67-3
IUPAC Namepyren-1-amine
Molecular FormulaC16H11N
InChI KeyYZVWKHVRBDQPMQ-UHFFFAOYSA-N
SMILESNC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)> 96.0%
Appearance (Color)Yellow to green
Assay (Titration)> 96.0%
FormPowder
It acts as a precursor molecule for production of other fluorescent probes such as 1-Azidopyrene, APTS, and N-(1-Pyrene)iodoacetamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only