Thermo Scientific Chemicals

Poly(methyl methacrylate), 600 micron, Thermo Scientific Chemicals

Catalog number: 043969.30
250 g, Each
Thermo Scientific Chemicals

Poly(methyl methacrylate), 600 micron, Thermo Scientific Chemicals

Catalog number: 043969.30
250 g, Each
Quantity
Catalog number: 043969.30
also known as 043969-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9011-14-7
Molecular Formula
(C5H8O2)n
SMILES
COC(=O)C(C)(-*)C-*
MDL Number
MFCD00134349
Appearance (Color)
White
Form
granular or spherical powder
Particle Size
600 micron

Description

Poly(methyl methacrylate), 600 microns is used in many medical and biochemical applications. It is suitable for 2D and 3D particle assemblies and for investigations in the colloidal crystal field due to the high monodispersity. It is also used in the field of one- and two-component toners for copying machines. Further, it is having a reduced non-specific protein binding activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text