Thermo Scientific Chemicals

5-Ethylidene-2-norbornene, endo + exo, 98%, stab. with 100ppm 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: 043972.AP
500 mL, Each
Thermo Scientific Chemicals

5-Ethylidene-2-norbornene, endo + exo, 98%, stab. with 100ppm 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: 043972.AP
500 mL, Each
Quantity
Catalog number: 043972.AP
also known as 043972-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16219-75-3
IUPAC Name
(5E)-5-ethylidenebicyclo[2.2.1]hept-2-ene
Molecular Formula
C9H12
InChI Key
OJOWICOBYCXEKR-KRXBUXKQNA-N
SMILES
CC=C1/CC2CC1C=C2
Appearance (Color)
Colorless
Form
liquid
Concentration
100 to 200 ppm BHT
Assay (GC)
> 97.5% ( sum of endo+exo isomers)

Description

5-Ethylidene-2-norbornene is used as a self healing agent for polymeric composites. It is also used to reinforce multiwalled carbon nanotubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text