Copper rod, 2.0mm (0.08in) dia, 99.99% (metals basis), Thermo Scientific Chemicals
Copper rod, 2.0mm (0.08in) dia, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper rod, 2.0mm (0.08in) dia, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
30 cm
10 cm
Catalog number 043973.BV
also known as 043973-BV
Price (USD)/ Each
69.65
Online exclusive
77.50 
Save 7.85 (10%)
-
Add to cart
Quantity:
30 cm
Request bulk or custom format
Price (USD)/ Each
69.65
Online exclusive
77.50 
Save 7.85 (10%)
Add to cart
Copper rod, 2.0mm (0.08in) dia, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number043973.BV
Price (USD)/ Each
69.65
Online exclusive
77.50 
Save 7.85 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.99% (metals basis)
FormRod, 2.0mm (0.08in) diameter.
Appearance (Color)Brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only