Thermo Scientific Chemicals
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99%, Thermo Scientific Chemicals
Catalog number: 043993.02
0.5 g, Each
Thermo Scientific Chemicals
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99%, Thermo Scientific Chemicals
Catalog number: 043993.02
0.5 g, Each
Quantity
Catalog number: 043993.02
also known as 043993-02
Price (USD)
Quantity
-
Have Questions?
Description
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III) is used precursor for aerosol-assisted CVD of mixed-conducting ceramic films of Sr-Co-Fe perovskite phases.