Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99%, Thermo Scientific Chemicals
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99%, Thermo Scientific Chemicals

Vapor deposition
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
0.5 g
10 g
Catalog number 043993.04
also known as 043993-04
Price (USD)/ Each
188.00
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
188.00
Add to cart
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99%, Thermo Scientific Chemicals
Catalog number043993.04
Price (USD)/ Each
188.00
-
Add to cart
Specifications
Chemical Name or MaterialTris (2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt (III)
CAS14877-41-9
Health Hazard 1H317-H351
Health Hazard 2GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
View more
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III) is used precursor for aerosol-assisted CVD of mixed-conducting ceramic films of Sr-Co-Fe perovskite phases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only