N,N-Dimethylformamide, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
N,N-Dimethylformamide, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Dimethylformamide, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 x 1 L
Catalog number 043997.K2
also known as 043997-K2
Price (USD)/ Each
157.65
Online exclusive
175.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
157.65
Online exclusive
175.00 
Save 17.35 (10%)
Add to cart
N,N-Dimethylformamide, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Catalog number043997.K2
Price (USD)/ Each
157.65
Online exclusive
175.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS68-12-2
IUPAC NameN,N-dimethylformamide
Molecular FormulaC3H7NO
InChI KeyZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILESCN(C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on Evaporation≤5 ppm
Assay (GC)≥99.8% (Corrected for water)
Water Content (Karl Fischer Titration)≤50 ppm
FormLiquid
Appearance (Color)Colorless
View more
N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only