Thermo Scientific Chemicals

Dimethyl sulfoxide, anhydrous, 99.8+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 043998.K2
1 L, Each
Thermo Scientific Chemicals

Dimethyl sulfoxide, anhydrous, 99.8+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 043998.K2
1 L, Each
Quantity
Catalog number: 043998.K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-68-5
IUPAC Name
methanesulfinylmethane
Molecular Formula
C2H6OS
InChI Key
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILES
CS(C)=O
Form
Liquid
Appearance (Color)
Colorless
Assay (GC)
≥99.8% (Corrected for water)
Residue on Evaporation
≤5 ppm
Comment
Inert gas used can be either Argon or Nitrogen

Description

Dimethyl sulfoxide is used as a solvent for chemical reactions involving salts, most notably in Finkelstein reactions and in other nucleophilic substitutions. DMSO is an effective pain relieving agent. Used in antifreeze or hydraulic fluids. Utilized in the oxidation of thiols and disulfides to sulfonic acids. Used to enhance dermal absorption of many chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text