Thermo Scientific Chemicals

Tetra-n-butylammonium perchlorate, electrochemical grade, Thermo Scientific Chemicals

Catalog number: 043999.18
50 g, Each
Thermo Scientific Chemicals

Tetra-n-butylammonium perchlorate, electrochemical grade, Thermo Scientific Chemicals

Catalog number: 043999.18
50 g, Each
Quantity
Catalog number: 043999.18
also known as 043999-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1923-70-2
IUPAC Name
tetrabutylazanium perchlorate
Molecular Formula
C16H36ClNO4
InChI Key
KBLZDCFTQSIIOH-UHFFFAOYSA-M
SMILES
[O-][Cl](=O)(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
Appearance (Color)
White
Form
Crystalline solid
Assay from Supplier's CofA
>98%

Description

Suitable for use as a polarographic supporting electrolyteTetra-n-butylammonium perchlorate is used as a supporting electrolyte in polarographic measurements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text