Tetra-n-butylammonium perchlorate, electrochemical grade, Thermo Scientific Chemicals
Tetra-n-butylammonium perchlorate, electrochemical grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetra-n-butylammonium perchlorate, electrochemical grade, Thermo Scientific Chemicals

Suitable for use as a polarographic supporting electrolyte
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number 043999.30
also known as 043999-30
Price (USD)/ Each
606.65
Online exclusive
674.00 
Save 67.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
606.65
Online exclusive
674.00 
Save 67.35 (10%)
Add to cart
Tetra-n-butylammonium perchlorate, electrochemical grade, Thermo Scientific Chemicals
Catalog number043999.30
Price (USD)/ Each
606.65
Online exclusive
674.00 
Save 67.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1923-70-2
IUPAC Nametetrabutylazanium perchlorate
Molecular FormulaC16H36ClNO4
InChI KeyKBLZDCFTQSIIOH-UHFFFAOYSA-M
SMILES[O-][Cl](=O)(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline solid
Assay from Supplier's CofA>98%
Suitable for use as a polarographic supporting electrolyteTetra-n-butylammonium perchlorate is used as a supporting electrolyte in polarographic measurements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only