Diethyl phosphite, 97+%, Thermo Scientific Chemicals
Diethyl phosphite, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl phosphite, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044000.14
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS762-04-9
IUPAC Namediethoxy(oxo)-λ⁵-phosphanylium
Molecular FormulaC4H10O3P
InChI KeyLXCYSACZTOKNNS-UHFFFAOYSA-N
SMILESCCO[P+](=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
FormLiquid
Assay (GC)≥97.0%
GC/MS AnalysisConform
Diethyl phosphite is precursor used for the synthesis of various organophosphorus compounds. used to prepare the nickel chloride-diethyl phosphite system, efficient catalyst for the cross-coupling reaction between various functionalized arylzinc halides and aryl bromides, triflates and activated chlorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only