Thermo Scientific Chemicals

Diethyl phosphite, 97+%, Thermo Scientific Chemicals

Catalog number: 044000.36
500 g, Each
Thermo Scientific Chemicals

Diethyl phosphite, 97+%, Thermo Scientific Chemicals

Catalog number: 044000.36
500 g, Each
Quantity
Catalog number: 044000.36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
762-04-9
IUPAC Name
diethoxy(oxo)-λ⁵-phosphanylium
Molecular Formula
C4H10O3P
InChI Key
LXCYSACZTOKNNS-UHFFFAOYSA-N
SMILES
CCO[P+](=O)OCC
Appearance (Color)
Clear, colorless
Form
Liquid
Assay (GC)
≥97.0%
GC/MS Analysis
Conform

Description

Diethyl phosphite is precursor used for the synthesis of various organophosphorus compounds. used to prepare the nickel chloride-diethyl phosphite system, efficient catalyst for the cross-coupling reaction between various functionalized arylzinc halides and aryl bromides, triflates and activated chlorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text