Thermo Scientific Chemicals

Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, Thermo Scientific Chemicals

Catalog number: 044031.06
5 g, Each
Thermo Scientific Chemicals

Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, Thermo Scientific Chemicals

Catalog number: 044031.06
5 g, Each
Quantity
Catalog number: 044031.06
also known as 044031-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
35138-22-8
IUPAC Name
λ¹-rhodium(1+) bis((1Z,5Z)-cycloocta-1,5-diene) tetrafluoroboranuide
Molecular Formula
C16H24BF4Rh
InChI Key
NBGSCPNNKGVTKU-QMDOQEJBSA-N
SMILES
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Assay (unspecified)
Rh 24.30-26.30%

Description

Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate is useful for hydrogenation, isomerization reactions and is also employed as a precursor for asymmetric hydrogenations. Furthermore, it is used in the synthesis of cationic COD-Rhodium complexes with phosphine ligands. In addition, it shows an important application for enantioselective hydrogenation and hydrosilylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text