Diphenyl(1,5-cyclooctadiene)platinum(II), Thermo Scientific Chemicals
Diphenyl(1,5-cyclooctadiene)platinum(II), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diphenyl(1,5-cyclooctadiene)platinum(II), Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number 044039.03
also known as 044039-03
Price (USD)/ Each
399.65
Online exclusive
444.00 
Save 44.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
399.65
Online exclusive
444.00 
Save 44.35 (10%)
Add to cart
Diphenyl(1,5-cyclooctadiene)platinum(II), Thermo Scientific Chemicals
Catalog number044039.03
Price (USD)/ Each
399.65
Online exclusive
444.00 
Save 44.35 (10%)
-
Add to cart
Chemical Identifiers
CAS12277-88-2
IUPAC Name(1Z,5Z)-cycloocta-1,5-diene; diphenylplatinumbis(ylium)
Molecular FormulaC20H22Pt
InChI KeyMQCNDJYHICNUPD-PHFPKPIQSA-N
SMILESC1C\C=C/CC\C=C/1.[Pt++](C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only