Thermo Scientific Chemicals

Yttrium(III) oxide, REacton™, nanopowder, 99.995% (REO), Thermo Scientific Chemicals

Catalog number: 044048.36
500 g, Each
Thermo Scientific Chemicals

Yttrium(III) oxide, REacton™, nanopowder, 99.995% (REO), Thermo Scientific Chemicals

Catalog number: 044048.36
500 g, Each
Quantity
Catalog number: 044048.36
also known as 044048-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1314-36-9
IUPAC Name
diyttrium(3+) trioxidandiide
Molecular Formula
O3Y2
InChI Key
RUDFQVOCFDJEEF-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Y+3].[Y+3]
Total Rare Earth Oxide Impurities
0.005% max.

Description

Yttriium(III) oxide is used to make phosphors for color TV tubes, yttrium iron garnets for use as microwave filters, high temperature superconductors, as a mineralizing agent for the formation of spinel and as a stabilizer for high-temperature service materials. It is presurcor for important inorganic compounds. It is a potential solid-state laser material which allows the efficient operation both in continuous operation and in pulsed regimes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text