Neodymium rod, 6.35mm (0.25in) dia, 99.5% (metals basis excluding Ta), Thermo Scientific Chemicals
Neodymium rod, 6.35mm (0.25in) dia, 99.5% (metals basis excluding Ta), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Neodymium rod, 6.35mm (0.25in) dia, 99.5% (metals basis excluding Ta), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mm
50 mm
100 mm
Catalog number 044051.HF
also known as 044051-HF
Price (USD)/ Each
128.00
-
Add to cart
Quantity:
10 mm
Request bulk or custom format
Price (USD)/ Each
128.00
Add to cart
Neodymium rod, 6.35mm (0.25in) dia, 99.5% (metals basis excluding Ta), Thermo Scientific Chemicals
Catalog number044051.HF
Price (USD)/ Each
128.00
-
Add to cart
Chemical Identifiers
CAS7440-00-8
IUPAC Nameneodymium
Molecular FormulaNd
InChI KeyQEFYFXOXNSNQGX-UHFFFAOYSA-N
SMILES[Nd]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities excluding Ta0.5% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged in mylar under argon
RUO – Research Use Only