Thermo Scientific Chemicals

Europium(III) oxide, REacton™, nanopowder, 99.999% (REO), Thermo Scientific Chemicals

Catalog number: 044052.14
25 g, Each
Thermo Scientific Chemicals

Europium(III) oxide, REacton™, nanopowder, 99.999% (REO), Thermo Scientific Chemicals

Catalog number: 044052.14
25 g, Each
Quantity
Catalog number: 044052.14
also known as 044052-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1308-96-9
IUPAC Name
dieuropium(3+) trioxidandiide
Molecular Formula
Eu2O3
InChI Key
AEBZCFFCDTZXHP-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Eu+3].[Eu+3]
Total Rare Earth Oxide Impurities
0.001% max.

Description

Europium(III) oxide is used as an agent for the manufacture of fluorescent glass. It can be used in the anti-counterfeiting phosphors in euro banknotes, red or blue phosphor in television sets. It shows a useful application as an activator for yttrium based phosphors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text