Thermo Scientific Chemicals

Ammonium tetrafluoroborate, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 044055.06
5 g, Each
Thermo Scientific Chemicals

Ammonium tetrafluoroborate, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 044055.06
5 g, Each
Quantity
Catalog number: 044055.06
also known as 044055-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13826-83-0
IUPAC Name
ammonium tetrafluoroboranuide
Molecular Formula
BF4H4N
InChI Key
PDTKOBRZPAIMRD-UHFFFAOYSA-O
SMILES
[NH4+].F[B-](F)(F)F
Total Metal Impurities
0.001% max.

Description

Ammonium fluoroborate is used as an analytical reagent. It is also applied in the textile printing and dyeing industry and used as a catalyst for resin finishing. It is mainly used for the high temperature fluxing action required by the metals industry; catalyst; in flame retardants and acts as a solid lubricant in cutting-oil emulsions for aluminum rolling and forming.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text