Thermo Scientific Chemicals

Tetraethylammonium perchlorate, 98% (dry wt.), cont. ca 10% water, Thermo Scientific Chemicals

Catalog number: 044062.09
10 g, Each
Thermo Scientific Chemicals

Tetraethylammonium perchlorate, 98% (dry wt.), cont. ca 10% water, Thermo Scientific Chemicals

Catalog number: 044062.09
10 g, Each
Quantity
Catalog number: 044062.09
also known as 044062-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2567-83-1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay from Supplier's CofA
≥97.5% (dry weight basis)
Assay from Supplier's CofA
Water content: ≥10.0 to ≤12.0%

Description

Tetraethylammonium perchlorate is used as supporting electrolyte in polarographic measurements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text