1-Methyl-2-pyrrolidinone, Biograde, 99.5%, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, Biograde, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, Biograde, 99.5%, Thermo Scientific Chemicals

Suitable for Biotech applications.Useful for spectrophotometry, chromatography and ICP-MS detection. Also, used for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide.
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
4 x 4 L
Packaging:
Under Argon
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044063.K7
also known as 044063-K7
Price (USD)/ Each
264.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
311.00 
Save 46.35 (15%)
-
Add to cart
Quantity:
4 L
Packaging:
Under Argon
Request bulk or custom format
Price (USD)/ Each
264.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
311.00 
Save 46.35 (15%)
Add to cart
1-Methyl-2-pyrrolidinone, Biograde, 99.5%, Thermo Scientific Chemicals
Catalog number044063.K7
Price (USD)/ Each
264.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
311.00 
Save 46.35 (15%)
-
Add to cart
Chemical Identifiers
CAS872-50-4
IUPAC Name1-methylpyrrolidin-2-one
Molecular FormulaC5H9NO
InChI KeySECXISVLQFMRJM-UHFFFAOYSA-N
SMILESCN1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofAAmines (as Dimethylamine) ≤10.0 ppm
Specific Absorbance (E 1%/1cm)285 nm ≤ 1.00 Au
Specific Absorbance (E 1%/1cm)325 nm ≤ 0.10 Au
Appearance (Color)Colorless, ≤20 APHA
Assay (GC)GC-FID ≥99.5%
View more
Suitable for Biotech applications.1-methyl-2-pyrrolidinone, is useful for spectrophotometry, chromatography and ICP-MS detection. It is useful for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. It is also useful for pharmaceutical and different industrial applications due to its broad solvency and low volatility.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Suitable for Biotech applications
  • Useful for spectrophotometry, chromatography and ICP-MS detection
  • Used for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide
  • Also useful for pharmaceutical, different industrial applications due to its broad solvency and low volatility
     
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only