Silver plating solution, metal content ≈28.7g/L, Thermo Scientific Chemicals
Silver plating solution, metal content ≈28.7g/L, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver plating solution, metal content ≈28.7g/L, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 L
Catalog number 044067.K2
also known as 044067-K2
Price (USD)/ Each
148.00
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
148.00
Add to cart
Silver plating solution, metal content ≈28.7g/L, Thermo Scientific Chemicals
Catalog number044067.K2
Price (USD)/ Each
148.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialSilver plating solution
Name NoteMetal content ≈28.7g/L
ColorColorless
Health Hazard 1H300-H310-H330-H400-H410: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
DOT InformationDOT Class: 6.1, Packaging Group: II
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only