Poly(vinylidene fluoride), Thermo Scientific Chemicals
Poly(vinylidene fluoride), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(vinylidene fluoride), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
500 g
2 kg
Catalog number 044080.A9
also known as 044080-A9
Price (USD)/ Each
956.65
Online exclusive
1063.00 
Save 106.35 (10%)
-
Add to cart
Quantity:
10 kg
Request bulk or custom format
Price (USD)/ Each
956.65
Online exclusive
1063.00 
Save 106.35 (10%)
Add to cart
Poly(vinylidene fluoride), Thermo Scientific Chemicals
Catalog number044080.A9
Price (USD)/ Each
956.65
Online exclusive
1063.00 
Save 106.35 (10%)
-
Add to cart
Chemical Identifiers
CAS24937-79-9
Molecular Formula(C2H2F2)n
SMILESFC(F)(-*)C-*
MDL NumberMFCD00084470
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
ViscosityMelt viscosity 23.50 to 29.50 Kpoise
FormPowder
Poly(vinylidene fluoride) is useful in coatings, film, filter cloth, instrument linings, filtration membranes, pump parts, linings and valves. It is also employed as a material for transducers in devices such as headphones, microphones and sonic detectors. It is also essential for piezoelectric and electrostrictive applications. In addition, it is useful for monofilament fishing lines replacing nylon monofilament. It acts as a standard binder used in the production of composite electrodes for lithium ion batteries. It serves as a high grade insulator for wires, nanomaterials and pressure sensors. It is utilized to make filters which are used to prepare samples for high performance liquid chromatography and other advanced analytical techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only