Thermo Scientific Chemicals

Chromium(III) potassium sulfate dodecahydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 044083.22
100 g, Each
Thermo Scientific Chemicals

Chromium(III) potassium sulfate dodecahydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 044083.22
100 g, Each
Quantity
Catalog number: 044083.22
also known as 044083-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7788-99-0
IUPAC Name
chromium(3+) potassium dodecahydrate disulfate
Molecular Formula
CrH24KO20S2
InChI Key
ZFVHBEKVAITXHW-UHFFFAOYSA-J
SMILES
O.O.O.O.O.O.O.O.O.O.O.O.[K+].[Cr+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Total Metal Impurities
0.1% max.

Description

It is used for tanning of leather and as a photographic film hardener. It is also used in textile dye, photography, ceramics, and the preparation of chromium standard solutions, as a catalyst, mordant, ceramics, tannage and developer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text