Thermo Scientific Chemicals

Boron nitride, Refractory Brushable Paint, BN 31%, Thermo Scientific Chemicals

Catalog number: 044086.A1
1 kg, Each
Thermo Scientific Chemicals

Boron nitride, Refractory Brushable Paint, BN 31%, Thermo Scientific Chemicals

Catalog number: 044086.A1
1 kg, Each
Quantity
Catalog number: 044086.A1
also known as 044086-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10043-11-5
IUPAC Name
boranimine
Molecular Formula
BN
InChI Key
LNLSXDSWJBUPHM-UHFFFAOYSA-N
SMILES
B=N
Form
Paste
pH
7-10
Assay from Supplier's CofA
Solids: 31.0-36.0%
Viscosity
3000-12000 cps

Description

Boron nitride is used as a lubricant and an additive to cosmetic products. It is also used in parts of high-temperature equipment, nanotechnology, polymorph. It is also used in used in xerographic process and laser printers. It is also used in fiberglass, ceramics, wood preservative, nuclear application and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text