Thermo Scientific Chemicals

1-Naphthylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 044088.09
10 g, Each
Thermo Scientific Chemicals

1-Naphthylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 044088.09
10 g, Each
Quantity
Catalog number: 044088.09
also known as 044088-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
552-46-5
IUPAC Name
hydrogen naphthalen-1-amine chloride
Molecular Formula
C10H10ClN
InChI Key
FOKKJVHTXPJHEN-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC1=C2C=CC=CC2=CC=C1
Appearance (Color)
white to grey
Assay (Titration)
> 97.5%
Form
powder

Description

Applied as a synthesis reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text