Thermo Scientific Chemicals

Silica gel, spherical, 300 angstroms, Thermo Scientific Chemicals

Catalog number: 044101.18
50 g, Each
Thermo Scientific Chemicals

Silica gel, spherical, 300 angstroms, Thermo Scientific Chemicals

Catalog number: 044101.18
50 g, Each
Quantity
Catalog number: 044101.18
also known as 044101-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7631-86-9
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Particle Size
d10/d90: ≤1.60 µm
Particle Size
d50: 18-22 µm
Particle Size
Pore Volume 0.7-0.9 mL/g
Surface Area
Report
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text