Thermo Scientific Chemicals

n-Hexylphosphonic acid, Thermo Scientific Chemicals

Catalog number: 044121.09
10 g, Each
Thermo Scientific Chemicals

n-Hexylphosphonic acid, Thermo Scientific Chemicals

Catalog number: 044121.09
10 g, Each
Quantity
Catalog number: 044121.09
also known as 044121-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4721-24-8
IUPAC Name
hexylphosphonate
Molecular Formula
C6H13O3P
InChI Key
GJWAEWLHSDGBGG-UHFFFAOYSA-L
SMILES
CCCCCCP([O-])([O-])=O
Purity
≥97.0%
Appearance (Color)
White to off-white
Form
Powder

Description

n-Hexylphosphonic acid is used in the manufacturing of nanoparticles such as quantum dots, nano-metals and nano-ceramics. It is also used for coating of many materials, including nanoparticles by condensed hydrophobic monolayers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text