Thermo Scientific Chemicals

Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride, Thermo Scientific Chemicals

Catalog number: 044123.02
0.5 g, Each
Thermo Scientific Chemicals

Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride, Thermo Scientific Chemicals

Catalog number: 044123.02
0.5 g, Each
Quantity
Catalog number: 044123.02
also known as 044123-02
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
36309-88-3
Form
Crystalline powder
Appearance (Color)
Orange to orange-red

Description

Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride is used as a probe for luminescent detection and quantitation of oxygen. It is also used for fiber optic sensors. Further, it is used in studies on oxygen in skin and skin tumors as well as measurement of oxygen flux through the skin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text