Rhodium 10%, Palladium 1% on activated carbon powder, Thermo Scientific Chemicals
Rhodium 10%, Palladium 1% on activated carbon powder, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rhodium 10%, Palladium 1% on activated carbon powder, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Catalog number 044124.14
also known as 044124-14
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialRhodium
Name Note10%, Palladium 1% on activated carbon powder
CAS7440-16-6
Health Hazard 1H228
Health Hazard 2GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only