beta-Nicotinamide adenine dinucleotide phosphate monosodium salt, Thermo Scientific Chemicals
beta-Nicotinamide adenine dinucleotide phosphate monosodium salt, Thermo Scientific Chemicals
Thermo Scientific Chemicals

beta-Nicotinamide adenine dinucleotide phosphate monosodium salt, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number 044126.03
also known as 044126-03
Price (USD)/ Each
327.65
Online exclusive
364.00 
Save 36.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
327.65
Online exclusive
364.00 
Save 36.35 (10%)
Add to cart
beta-Nicotinamide adenine dinucleotide phosphate monosodium salt, Thermo Scientific Chemicals
Catalog number044126.03
Price (USD)/ Each
327.65
Online exclusive
364.00 
Save 36.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1184-16-3
IUPAC Namesodium 1-[(2R,3R,4S,5R)-5-({[({[(2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonooxy)oxolan-2-yl]methyl phosphonato}oxy)(oxido)phosphoryl]oxy}methyl)-3,4-dihydroxyoxolan-2-yl]-3-carbamoyl-1λ⁵-pyridin-1-ylium
Molecular FormulaC21H27N7NaO17P3
InChI KeyJNUMDLCHLVUHFS-QYZPTAICSA-M
SMILES[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Assay (special)Sodium: 1.5-4.5%
Purity≥97.0%
FormPowder
β-Nicotinamide adenine dinucleotide phosphate monosodium salt acts as an electron acceptor used by cells. It is also useful in vitro to study NADP. Furthermore, it is used in a wide range of enzyme catalyzed oxidation reduction reactions. The combination of NADP+/NADPH is employed in a variety of antioxidation mechanisms, which protect against reactive oxidation species accumulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only