Nickel gauze, 100 mesh woven from 0.1mm (0.004in) dia wire, Thermo Scientific Chemicals
Nickel gauze, 100 mesh woven from 0.1mm (0.004in) dia wire, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel gauze, 100 mesh woven from 0.1mm (0.004in) dia wire, Thermo Scientific Chemicals

Form: Wire Cloth; Open area: 36%; Width of opening: 0.006 in.
Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
Catalog number 044128.FL
also known as 044128-FL
Price (USD)/ Each
71.65
Online exclusive
79.80 
Save 8.15 (10%)
-
Add to cart
Quantity:
75 x 75 mm
Request bulk or custom format
Price (USD)/ Each
71.65
Online exclusive
79.80 
Save 8.15 (10%)
Add to cart
Nickel gauze, 100 mesh woven from 0.1mm (0.004in) dia wire, Thermo Scientific Chemicals
Catalog number044128.FL
Price (USD)/ Each
71.65
Online exclusive
79.80 
Save 8.15 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentNo Specifications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only