Thermo Scientific Chemicals

Lithium methoxide, 98+%, Thermo Scientific Chemicals

Catalog number: 044129.14
25 g, Each
Thermo Scientific Chemicals

Lithium methoxide, 98+%, Thermo Scientific Chemicals

Catalog number: 044129.14
25 g, Each
Quantity
Catalog number: 044129.14
also known as 044129-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
865-34-9
Assay (unspecified)
98.0% min

Description

Lithium methoxide is used in sol-gel synthesis of lithium borophosphates. It is also useful for deprotonation reactions in organic synthesis. Further, it serves as a mild base in organic synthesis especially in transesterifications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text