N,N'-Bis(acryloyl)cystamine, 98%, Thermo Scientific Chemicals
N,N'-Bis(acryloyl)cystamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N'-Bis(acryloyl)cystamine, 98%, Thermo Scientific Chemicals

Used as a reversible cross-linker for polyacrylamide gels
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 044132.03
also known as 044132-03
Price (USD)/ Each
69.65
Online exclusive
77.30 
Save 7.65 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
69.65
Online exclusive
77.30 
Save 7.65 (10%)
Add to cart
N,N'-Bis(acryloyl)cystamine, 98%, Thermo Scientific Chemicals
Catalog number044132.03
Price (USD)/ Each
69.65
Online exclusive
77.30 
Save 7.65 (10%)
-
Add to cart
Chemical Identifiers
CAS60984-57-8
IUPAC NameN-(2-{[2-(prop-2-enamido)ethyl]disulfanyl}ethyl)prop-2-enamide
Molecular FormulaC10H16N2O2S2
InChI KeyDJVKJGIZQFBFGS-UHFFFAOYSA-N
SMILESC=CC(=O)NCCSSCCNC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥97.5%
Melting Point121-126°C
Appearance (Color)White
FormPowder
N,N'-Bis(acryloyl)cystamine is used as a reversible cross-linker for polyacrylamide gels. It is also used for DNA purification. Further, it finds application in Suzuki reaction and fabrication of hydrogel modified plasmonic crystal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only