Lithium tert-butoxide, 99.9% (metals basis), Thermo Scientific Chemicals
Lithium tert-butoxide, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific™

Lithium tert-butoxide, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 044133.04
also known as 044133-04
Price (USD)/ Each
24.65
Online exclusive
27.60 
Save 2.95 (11%)
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
24.65
Online exclusive
27.60 
Save 2.95 (11%)
Add to cart
Lithium tert-butoxide, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number044133.04
Price (USD)/ Each
24.65
Online exclusive
27.60 
Save 2.95 (11%)
-
Add to cart
Chemical Identifiers
CAS1907-33-1
IUPAC Namelithium(1+) 2-methylpropan-2-olate
Molecular FormulaC4H9LiO
InChI KeyLZWQNOHZMQIFBX-UHFFFAOYSA-N
SMILES[Li+].CC(C)(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride content≤300ppm
Trace MetalBa: ≤1ppm
Trace MetalMg: ≤1ppm
Trace MetalCa: ≤200ppm
FormPowder
View more

Lithium tert-butoxide is a mild base used in organic synthesis. It is involved in the alfa-alkylation reaction of ketones with primary alcohols. It is utilized as a catalyst in the reduction of ketones to the corresponding secondary alcohols. It acts as lithium source utilized in the preparation of LixTiyOz thin films. Further, it is used in conjunction with organometallic reagents/catalysts to modify their reactivity and selectivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only