Thermo Scientific Chemicals

MOPSO, 98%, Thermo Scientific Chemicals

Catalog number: 044135.A1
1 kg, Each
Thermo Scientific Chemicals

MOPSO, 98%, Thermo Scientific Chemicals

Catalog number: 044135.A1
1 kg, Each
Quantity
Catalog number: 044135.A1
also known as 044135-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68399-77-9
IUPAC Name
4-[(2S)-2-hydroxy-3-sulfonatopropyl]morpholin-4-ium
Molecular Formula
C7H15NO5S
InChI Key
NUFBIAUZAMHTSP-ZETCQYMHSA-N
SMILES
O[C@@H](C[NH+]1CCOCC1)CS([O-])(=O)=O
Loss on Drying
≤1.0%
Appearance (Color)
White
Assay from Supplier's CofA
≥97.5%
Solution Test
Clear and complete (10% in water)
Form
Powder

Description

It is an important raw material used intermediate used in pharmaceuticals, organic synthesis, dye stuffs and agrochemicals. Also used as biological buffers, pH range 6.2-7.6.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text