Silicon(IV) iodide, 99.999% (metals basis), Thermo Scientific Chemicals
Silicon(IV) iodide, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silicon(IV) iodide, 99.999% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number 044141.14
also known as 044141-14
Price (USD)/ Each
551.65
Online exclusive
613.00 
Save 61.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
551.65
Online exclusive
613.00 
Save 61.35 (10%)
Add to cart
Silicon(IV) iodide, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number044141.14
Price (USD)/ Each
551.65
Online exclusive
613.00 
Save 61.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13465-84-4
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Silicon(IV) iodide is used as a precursor to prepare silicon amides. It finds application in the manufacture and etching of silicon in microelectronics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only