1x1 image pixel for data collection
Palladium, 5% on activated carbon powder, standard, reduced, acidic catalyst, nominally 50% water wet
Palladium, 5% on activated carbon powder, standard, reduced, acidic catalyst, nominally 50% water wet
Palladium, 5% on activated carbon powder, standard, reduced, acidic catalyst, nominally 50% water wet
Palladium, 5% on activated carbon powder, standard, reduced, acidic catalyst, nominally 50% water wet
Thermo Scientific Chemicals

Palladium, 5% on activated carbon powder, standard, reduced, acidic catalyst, nominally 50% water wet

CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 044142.18
also known as 044142-18
Price (USD)
-
Quantity:
50 g
Request bulk or custom format
Specifications
CAS7440-05-3
ChEBICHEBI:33363
Chemical Name or MaterialPalladium
IUPAC Namepalladium
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
View more
Ask our AI about this Product
Hydrogenation of aromatic and aliphatic nitro groups, reductive alkylation/amination, hydrogenation of aromatic carbonyls, olefins, nitriles

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Hydrogenation of aromatic and aliphatic nitro groups, reductive alkylation/amination, hydrogenation of aromatic carbonyls, olefins, nitriles
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
Q03J028Certificate of AnalysisMay 09, 2023044142.09
W25F037Certificate of AnalysisNov 22, 2021044142.18, 044142.30, 044142.09
I28R044Certificate of AnalysisNov 22, 2021044142.18, 044142.30, 044142.09
R07F033Certificate of AnalysisNov 22, 2021044142.18, 044142.30, 044142.09
L07S019Certificate of AnalysisNov 22, 2021044142.18, 044142.30, 044142.09
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection