Lithium dimethylamide, 96%, Thermo Scientific Chemicals
Lithium dimethylamide, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium dimethylamide, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 044145.06
also known as 044145-06
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
Add to cart
Lithium dimethylamide, 96%, Thermo Scientific Chemicals
Catalog number044145.06
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS3585-33-9
IUPAC Namelithium(1+) dimethylamine hydride
Molecular FormulaC2H8LiN
InChI KeyQHXBFEDDNKOZHB-UHFFFAOYSA-N
SMILES[H-].[Li+].CNC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormPowder and/or chunks
Assay from Supplier's CofA≥95.0% (titration)
It is used in the synthesis of some d0 organoimido complexes of the early transition metals and in meta rearrangement in the reaction of triphenyl-(p-bromophenyl)-silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only